Skip to Content
Merck
1-Iodo-2,2-dimethylpropane 98%

1-Iodo-2,2-dimethylpropane

1-Iodo-2,2-dimethylpropane 98%
Synonym(s):
2,2-Dimethyl-1-iodopropane, Neopentyl iodide
Linear Formula:
(CH3)3CCH2I
CAS Number:
Molecular Weight:
198.05
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing