Skip to Content
Merck
1-Chlorododecane ≥97.0% (GC)

1-Chlorododecane

1-Chlorododecane ≥97.0% (GC)
Synonym(s):
Dodecyl chloride, Lauryl chloride, 1-Chlorododecane
Linear Formula:
CH3(CH2)11Cl
CAS Number:
Molecular Weight:
204.78

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing