(1,5-Cyclooctadiene)(pyridine)(tricyclohexylphosphine)-iridium(I) hexafluorophosphate
Synonym(s):
(1,5-Cyclooctadiene)(pyridine)(tricyclohexylphosphine)-Ir(I) PF6, Crabtree’s catalyst, Iridium(I) hexafluorophosphate (1,5-Cyclooctadiene)-(pyridine)-(tricyclohexylphosphine) complex, [Ir(cod)(PCy3)(py)]PF6
Empirical Formula (Hill Notation):
C31H50F6IrNP2
CAS Number:
Molecular Weight:
804.89
Products
Brand
Formula Weight
Melting Point (°C)
Purity
Reagent Type
Core
Special Grade
Showing 1-2 of 2