1,3-Dimethyluric acid
Synonym(s):
1,3-Dimethyl-2,6,8-trihydroxypurine, 1,3-Dimethyl-7,9-dihydro-1H-purine-2,6,8(3H)-trione, Oxytheophylline
Empirical Formula (Hill Notation):
C7H8N4O3
CAS Number:
Molecular Weight:
196.16
Products
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Product Line
Format
Quality Segment
Application
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing