![1,2-Diacyl-sn-glycero-3-phospho-L-serine ≥97% (TLC), from bovine brain, amorphous powder](/deepweb/assets/sigmaaldrich/product/structures/767/597/0806b5b5-a047-4205-9418-f604d0de5ae6/640/0806b5b5-a047-4205-9418-f604d0de5ae6.png)
1,2-Diacyl-sn-glycero-3-phospho-L-serine
![1,2-Diacyl-sn-glycero-3-phospho-L-serine ≥97% (TLC), from bovine brain, amorphous powder](/deepweb/assets/sigmaaldrich/product/structures/767/597/0806b5b5-a047-4205-9418-f604d0de5ae6/640/0806b5b5-a047-4205-9418-f604d0de5ae6.png)
Synonym(s):
3-sn-Phosphatidyl-L-serine, L-α-Phosphatidyl-L-serine, PS, Phosphatidylserine
Products
Product Category
Brand
Biological Source
Physical Form
Purity
Tissue
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing