
10-desacetyl-baccatin III

Synonym(s):
10-Deacetylbaccatin III
Empirical Formula (Hill Notation):
C29H36O10
CAS Number:
Molecular Weight:
544.59
Products
Brand
Formula Weight
Agency
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing