Product Name
Ethyl 6-ethoxy-2-hydroxybenzoate 95%
Product Number
564974
Product Brand
aldrich
CAS Number
154364-61-1
Molecular Weight
210.23
Test | Specification |
---|---|
Appearance (Color) | White to Off-White |
Appearance (Form) | Conforms to Requirements |
Powder or Crystals | |
Infrared spectrum | Conforms to Structure |
Purity (GC) | ≥94.0 % |