Product Name
2,2,2-Trifluoro-2′,4′,6′-trimethylacetophenone ≥97%
Product Number
420476
Product Brand
aldrich
CAS Number
313-56-4
Molecular Weight
216.20
Test | Specification |
---|---|
APPEARANCE (COLOR) | Colorless to Yellow |
APPEARANCE (FORM) | Liquid |
PURITY (GC AREA %) | ≥ 97.0 % |
INFRARED SPECTRUM | CONFORMS TO STRUCTURE |