Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

55068

Peek adapter 10-32 TO M6, .020 in. hole

Synonym(s):

Tubing Adapter

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

UNSPSC Code:
41115700

material

PEEK adapter

description

for chromatography applications

packaging

pkg of 1 ea

parameter

1000 psi max. pressure (69 bar)

adapter swept volume

0.30 μL

bore diam.

0.020 in. (0.50 mm)

fitting

10-32 female for 1/16 in. O.D. tubing (fingertight fitting included)
M6 female

Looking for similar products? Visit Product Comparison Guide

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service