Skip to Content
Merck
All Photos(1)

Key Documents

40007

Supelco

1,2,4-Trichlorobenzene solution

certified reference material, 5000 μg/mL in methanol

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
41116105

grade

certified reference material
TraceCERT®

product line

TraceCERT®

CofA

current certificate can be downloaded

packaging

ampule of 1 mL

concentration

5000 μg/mL in methanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

single component solution

storage temp.

2-8°C

InChI

1S/C6H3Cl3/c7-4-1-2-5(8)6(9)3-4/h1-3H

InChI key

PBKONEOXTCPAFI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 2 - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jie Cao et al.
The Science of the total environment, 409(11), 2336-2341 (2011-03-29)
Monodispersed carboxymethyl cellulose (CMC)-stabilized Fe-Cu bimetal nanoparticles with an average diameter of less than 20nm were successfully synthesized by a modified water-based approach. The as-resulting particles exhibit a core-shell structure and are quite uniform in size and shape. Batch experiments
Jian Jiang et al.
Huan jing ke xue= Huanjing kexue, 29(6), 1655-1659 (2008-09-04)
Pseudomonas nitroreducens J5-1 is able to use monochlorobenzene, 1,2-dichlorobenzene, 1,3-dichlorobenzene and 1,2,4-trichlorobenzene as sole carbon and energy sources, and it differs from those 1,2,4-trichlorobenzene degrading bacteria reported in substrate utilizing characters. PCR technique was used to amplify the genes of
Tatyana Balandina et al.
Chemical communications (Cambridge, England), 49(22), 2207-2209 (2013-02-12)
STM brings to light chirality aspects of the self-assembly of a functionalized helicene at the interface between a liquid and the solid substrates, gold and graphite. This reveals conditions for conglomerate formation.
Ernest Marco-Urrea et al.
Bioresource technology, 100(23), 5757-5762 (2009-07-21)
Extracellular hydroxyl radical ((*)OH) production via quinone redox cycling in Trametes versicolor, grown in a chemically defined medium, was investigated to degrade trichloroethylene (TCE), perchloroethylene (PCE), 1,2,4- and 1,3,5-trichlorobenzene (TCB). The activity of the enzymes catalyzing the quinone redox cycle
Wei Zhang et al.
Huan jing ke xue= Huanjing kexue, 32(7), 1974-1979 (2011-09-20)
Effects and kinetics of photocatalytic degradation of 1,2,4-trichlorobenzene with composite photocatalysts of TiO2 coated on carbon nanotubes and nano-TiO2 photocatalysts were studied, respectively. It was found that the composite photocatalysts was better than the nano-TiO2 for photocatalytic degradation of 1,2,4-trichlorobenzene.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service