Skip to Content
Merck
All Photos(1)

Key Documents

Z380660

Millipore

Steriflip® centrifuge tube filter unit

pkg of 25 ea

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41103900
NACRES:
NB.24

material

polyethersulfone membrane

sterility

sterile

packaging

pkg of 25 ea

manufacturer/tradename

Millipore SCGP00525

tube size

50 mL

working volume

50 mL

pore size

0.22 μm

General description

Go from centrifuge to vacuum filtration with no sample transfer. Simply screw the unit to a 50 mL centrifuge tube, invert, and apply vacuum. The filtrate collects in another 50 mL centrifuge tube that is part of the Steriflip unit.

Membrane is Millipore Express low-protein binding polyethersulfone (PES).

Legal Information

Steriflip is a registered trademark of Merck KGaA, Darmstadt, Germany

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

  1. Which document(s) contains shelf-life or expiration date information for a given product?

    If available for a given product, the recommended re-test date or the expiration date can be found on the Certificate of Analysis.

  2. How do I get lot-specific information or a Certificate of Analysis?

    The lot specific COA document can be found by entering the lot number above under the "Documents" section.

  3. How do I find price and availability?

    There are several ways to find pricing and availability for our products. Once you log onto our website, you will find the price and availability displayed on the product detail page. You can contact any of our Customer Sales and Service offices to receive a quote.  USA customers:  1-800-325-3010 or view local office numbers.

  4. What is the Department of Transportation shipping information for this product?

    Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product. 

  5. Are the tubes of Product Z380660-25EA, Steriflip® centrifuge tube filter unit, individually wrapped?

    For Sigma-Aldrich Product Z380660-25EA (corresponding to Millipore Product SCGP00525), the tubes do come individually wrapped.

  6. My question is not addressed here, how can I contact Technical Service for assistance?

    Ask a Scientist here.

Renato Elias Rodrigues de Souza Santos et al.
Vector borne and zoonotic diseases (Larchmont, N.Y.), 20(2), 117-124 (2019-10-23)
Trypanosoma rangeli is an avirulent flagellate protozoan that could mislead correct diagnosis of Trypanosoma cruzi infection, the causative agent of Chagas' disease, given their high similarity. Besides, T. rangeli presents two genetic groups, whose differentiation is achieved mainly by molecular
Arianna Marini et al.
Frontiers in immunology, 10, 2931-2931 (2020-01-11)
Development of effective malaria vaccines requires delivery platforms to enhance the immunogenicity and efficacy of the target antigens. This is particularly challenging for transmission-blocking malaria vaccines (TBVs), and specifically for those based on the Pfs25 antigen, that need to elicit
Ruilian Ying et al.
Materials science & engineering. C, Materials for biological applications, 110, 110689-110689 (2020-03-25)
Hydroxyapatite/chitosan (HAp/CS) composites have been widely studied and applied in tissue engineering fields due to their excellent biocompatibility and degradability. However, to improve the mechanical properties of CS, cross-linking agents are commonly added, which will seriously affect its biocompatibility and
Jiří Skuhrovec et al.
Journal of economic entomology, 113(1), 255-262 (2019-11-07)
The Colorado potato beetle ranks as one of the most important potato pests, mainly due to its high feeding rate during all developmental stages, particularly third and fourth larval instar, and high fecundity. The effect of essential oil (EO) from
Mariana Avalos et al.
The ISME journal, 14(2), 569-583 (2019-11-09)
Soil-inhabiting streptomycetes are nature's medicine makers, producing over half of all known antibiotics and many other bioactive natural products. However, these bacteria also produce many volatiles, molecules that disperse through the soil matrix and may impact other (micro)organisms from a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service