Skip to Content
Merck
All Photos(2)

Key Documents

SRP5222

Sigma-Aldrich

MSK2, active, GST tagged from mouse

PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

RPS6KA4, RSK-B, RSKB

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

recombinant

expressed in baculovirus infected Sf9 cells

product line

PRECISIO® Kinase

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

7.5-10.4 nmol/min·mg

mol wt

~114 kDa

NCBI accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

mouse ... Rps6ka4(56613)

General description

MSK2 (mitogen- and stress-activated protein kinase-2) is a member of the serine/threonine kinase family that contains two non-identical kinase catalytic domains. MSK2 is also known as ribosomal protein S6 kinase polypeptide 4 (RPS6KA4), that is activated by the mitogen-activated protein kinases- extracellular signal-regulated kinases 1 and 2 (ERK1 and -2) and p38.

Biochem/physiol Actions

MSK2 (mitogen- and stress-activated protein kinase-2) phosphorylates various substrates like cyclic AMP-responsive element-binding protein 1 (CREB1) and the proto-oncogene c-fos. It is a histone-modifying enzyme and a mitogenic factor. The protein is involved in the G1-phase of the cell cycle and has a role in apoptosis. It also modulates the functions of various transcription factors.

Physical form

Supplied in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol.

Preparation Note

after opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Accelerated apoptotic death and in vivo turnover of erythrocytes in mice lacking functional mitogen- and stress-activated kinase MSK1/2.
Lang E
Scientific Reports, 5, 17316-17316 (2015)
ERK and p38 MAPK-activated protein kinases: a family of protein kinases with diverse biological functions.
Roux PP and Blenis J
Microbiology and Molecular Biology Reviews, 68(2), 320-344 (2004)
Mitogen and stress-activated kinases 1/2 regulate ischemia-induced hippocampal progenitor cell proliferation and neurogenesis.
Karelina K
Neuroscience, 285, 292-302 (2015)
Comparison of epigenetic mediator expression and function in mouse and human embryonic blastomeres.
Chavez SL, McElroy SL, Bossert NL, et al.
Human Molecular Genetics, 23(18), 4970-4984 (2014)
B Pierrat et al.
The Journal of biological chemistry, 273(45), 29661-29671 (1998-10-29)
A novel ribosomal S6 kinase (RSK) family member, RSK-B, was identified in a p38alphaMAPK-baited intracellular interaction screen. RSK-B presents two catalytic domains typical for the RSK family. The protein kinase C-like N-terminal and the calcium/calmodulin kinase-like C-terminal domains both contain

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service