Skip to Content
Merck
All Photos(1)

Key Documents

SML3235

Sigma-Aldrich

SG-005 Hydrochloride

≥98% (HPLC)

Synonym(s):

(±)-7-(Benzyloxy)-1-(4-(benzyloxy)benzyl)-6-methoxy-2-methyl-1,2,3,4-tetrahydro-isoquinoline Hydrochloride, 1,2,3,4-tetrahydro-6-methoxy-2-methyl-7-(phenylmethoxy)-1-[[4-(phenylmethoxy)phenyl]methyl]-isoquinoline hydrochloride (1:1)

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
831,00 kr.
25 MG
3.340,00 kr.

831,00 kr.


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
831,00 kr.
25 MG
3.340,00 kr.

About This Item

Empirical Formula (Hill Notation):
C32H33NO3 ·HCl
CAS Number:
Molecular Weight:
516.07
UNSPSC Code:
12352200
NACRES:
NA.77

831,00 kr.


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

Related Categories

Biochem/physiol Actions

SG-005, a tetrandrine derivative, is a potent and selective inhibitor of two-pore channel 2 (TPC2) that exhibits antiproliferative properties against cancer cells.
potent and selective inhibitor of two-pore channel 2 (TPC2) that exhibits antiproliferative properties against cancer cells

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service