Skip to Content
Merck
All Photos(2)

Key Documents

HPA008588

Sigma-Aldrich

Anti-NMI antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-N-myc and STAT interactor antibody produced in rabbit, Anti-N-myc-interactor antibody produced in rabbit, Anti-Nmi antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:20-1:50

immunogen sequence

KINVTEIPDTLREDQMRDKLELSFSKSRNGGGEVDRVDYDRQSGSAVITFVEIGVADKILKKKEYPLYINQTCHRVTVSPYTEIHLKKYQIFSGTSKRTVLLTGMEGIQMDEEIVEDLINIHFQRAKNGGGEVDVVKCSLGQPH

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NMI(9111)

General description

NMI (N-myc and STAT interactor) protein was initially identified in a yeast two-hybrid system, and is a homolog of interferon (IFN)-inducible protein IFP 35. It contains an Nmi/IFP 35 domain (NID), formed of 90-95 amino acids, and is responsible for NMI-NMI interactions and its localization within the cell. Its N-terminal contains the binding site for Myc-family proteins and other transcription factors. This protein is localized to human chromosome 22q13.3.

Immunogen

N-myc-interactor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

NMI (N-myc and STAT interactor) interacts with interferon (IFN)-inducible protein IFP 35 to form a protein complex, and thus, prevents the degration of IFP 35 by proteasome. Therefore, it amplifies the effects of IFNs by stabilizing IFP 35 protein. It acts as a tumor suppressor in breast cancer, where its expression is down-regulated. Up-regulation of this gene in aggressive breast cancer cell lines, results in inhibition of Wnt/β-catenin signaling, leading to retardation of tumor growth. It is also involved in the tumorigenesis of glioma, and variants of this gene are linked to susceptibility to gliomas.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST70910

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Delong Meng et al.
Tumour biology : the journal of the International Society for Oncodevelopmental Biology and Medicine, 36(3), 1579-1588 (2014-11-13)
Glioma is one of the most common and lethal brain tumors. N-myc (and STAT) interactor (NMI) gene has been reported in tumorigenesis, and our previous study further showed its implication in glioma progression. To elucidate its involvement in the etiology
X Zhou et al.
The Journal of biological chemistry, 275(28), 21364-21371 (2000-04-26)
Nmi and IFP35 are interferon (IFN)-induced proteins. In cells treated with IFN-gamma, Nmi enhances the association of transcription co-activator CBP/p300 with signal transducer and activator of transcription proteins, and IFP35 forms a high molecular weight cytosolic complex of unknown constituents.
Rebecca A Fillmore et al.
International journal of cancer, 125(3), 556-564 (2009-04-10)
We found that the expression levels of N-Myc interactor (Nmi) were low in aggressive breast cancer cell lines when compared with less aggressive cell lines. However, the lower levels in the aggressive lines were inducible by interferon-gamma (IFN-gamma). Because Nmi
J Chen et al.
The Journal of biological chemistry, 275(46), 36278-36284 (2000-08-22)
Nmi is an interferon (IFN)-inducible protein homologous to IFN-inducible protein IFP 35. The homology consists of a novel Nmi/IFP 35 domain (NID) of 90-92 amino acids that is repeated in tandem in each protein and mediates Nmi-Nmi protein interactions and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service