Skip to Content
Merck
All Photos(2)

Key Documents

E4890

Sigma-Aldrich

trans-9,10-Epoxystearic acid methyl ester

~98% (capillary GC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H36O3
CAS Number:
Molecular Weight:
312.49
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:

Assay

~98% (capillary GC)

form

solid

functional group

ester

shipped in

ambient

storage temp.

−20°C

SMILES string

CCCCCCCC[C@@H]1O[C@H]1CCCCCCCC(=O)OC

InChI

1S/C19H36O3/c1-3-4-5-6-8-11-14-17-18(22-17)15-12-9-7-10-13-16-19(20)21-2/h17-18H,3-16H2,1-2H3/t17-,18-/m0/s1

InChI key

CAMHHLOGFDZBBG-ROUUACIJSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Sevanian et al.
Biochimica et biophysica acta, 614(2), 489-500 (1980-08-07)
The activity of rat lung epoxide hydrolase (epoxide hydrolase, EC 3.3.2.3) was studied using two lipid epoxides which can be isolated from lung tissue. These epoxides displayed different Km,app and hydration rates. Methyl cis-9,10-epoxystearate was hydrated 20-times more rapidly than
C Fanelli et al.
Journal of general microbiology, 129(6), 1721-1723 (1983-06-01)
Epoxy fatty acids added to the culture media either with the inoculum or at the end of exponential growth phase stimulated aflatoxin production by toxigenic strains of Aspergillus flavus and Aspergillus parasiticus. This effect did not appear when the unsaturated
Atanu Biswas et al.
Journal of agricultural and food chemistry, 57(18), 8136-8141 (2009-09-02)
A facile (and environmentally friendly) reaction between epoxidized methyl oleate and aniline to produce an oleate-aniline adduct, without the formation of fatty amide, was discovered. This reaction was carried out neat, with a catalytic amount of an ionic liquid. No

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service