Key Documents
CLS3389
Corning® HTS Transwell® Micromatic 8 Channel aspirator
About This Item
Pricing and availability is not currently available.
description
Micromatic 8 channel aspirator
feature
autoclavable
manufacturer/tradename
Corning 3389
General description
Helps safely vacuum aspirate medium or buffers from the apical portion of the HTS Transwell-96 inserts. Using the 8-Channel Aspirator reduces cell death or detachment caused by too rapid removal of solutions or media during vacuum aspiration.
Legal Information
Corning is a registered trademark of Corning, Inc.
Transwell is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)