Skip to Content
Merck
All Photos(1)

Key Documents

A3825

Sigma-Aldrich

N-Acetylneuraminic acid methyl ester

≥98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H21NO9
CAS Number:
Molecular Weight:
323.30
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:

biological source

synthetic

Assay

≥98%

storage temp.

−20°C

SMILES string

O.COC(=O)C1(O)CC(O)C(NC(C)=O)C(O1)C(O)C(O)CO

InChI

1S/C12H21NO9.H2O/c1-5(15)13-8-6(16)3-12(20,11(19)21-2)22-10(8)9(18)7(17)4-14;/h6-10,14,16-18,20H,3-4H2,1-2H3,(H,13,15);1H2

InChI key

UIUNVNKHJXXRLV-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shoichi Nishitani et al.
ChemistryOpen, 7(7), 513-519 (2018-07-14)
The origin of the unusually high stability of the sialic acid (SA) and phenylboronic acid (PBA) complex was investigated by a combined nuclear magnetic resonance (NMR) spectroscopy and density functional theory (DFT) study. SA is a glycan-terminating monosaccharide, and its

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service