Skip to Content
Merck
All Photos(1)

Key Documents

84470

Sigma-Aldrich

Samarium(III) trifluoromethanesulfonate

purum, ≥97.0% (KT)

Synonym(s):

Samarium(III) triflate, Trifluoromethanesulfonic acid samarium(III) salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Sm(SO3CF3)3
CAS Number:
Molecular Weight:
597.57
MDL number:
UNSPSC Code:
12352300

grade

purum

Assay

≥97.0% (KT)

extent of labeling

26 specific activity

SMILES string

[Sm+3].[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F

InChI

1S/3CHF3O3S.Sm/c3*2-1(3,4)8(5,6)7;/h3*(H,5,6,7);/q;;;+3/p-3

InChI key

DDCWGUIPLGMBPO-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

Caution

radioactive

Other Notes

Effective Lewis acid catalyst; Used to prepare organo-samarium compounds

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S. Kobayashi et al.
Synthesis, 1195-1195 (1995)
S.-I. Fukuzawa et al
Chemistry Letters (Jpn), 1981-1981 (1994)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service