Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

46319

Supelco

3-Phenoxybenzoic acid

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

Linear Formula:
C6H5OC6H4CO2H
CAS Number:
Molecular Weight:
214.22
Beilstein:
2105574
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

147-149 °C (lit.)

application(s)

agriculture
environmental

format

neat

SMILES string

OC(=O)c1cccc(Oc2ccccc2)c1

InChI

1S/C13H10O3/c14-13(15)10-5-4-8-12(9-10)16-11-6-2-1-3-7-11/h1-9H,(H,14,15)

InChI key

NXTDJHZGHOFSQG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kateryna Babina et al.
Environment international, 48, 109-120 (2012-08-16)
Organophosphorus (OP) and pyrethroid (PYR) compounds are the most widely used insecticides. OPs and PYRs are developmental neurotoxicants. Understanding the extent of exposure in the general population and especially in young children is important for the development of public health
Hee-Joo Kim et al.
Analytical chemistry, 83(1), 246-253 (2010-12-15)
Immuno polymerase chain reaction (IPCR) is an analytical technology based on the excellent affinity and specificity of antibodies combined with the powerful signal amplification of polymerase chain reaction (PCR), providing superior sensitivity to classical immunoassays. Here we present a novel
C Fortes et al.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association, 52, 91-96 (2012-11-14)
Concerns about pesticide exposure through food consumption have increased during the past several years. Pyrethroids are applied as insecticides throughout the world. Human metabolism of pyrethroids results in urinary metabolites that are suitable for biological monitoring. The objective of our
Bartosz Wielgomas et al.
International journal of hygiene and environmental health, 216(3), 295-300 (2012-10-02)
Urinary concentrations of pyrethroid metabolites were measured in the first void urine samples collected from 132 healthy people living in the Gdańsk region of Northern Poland in 2010 and 2011. Four metabolites of synthetic pyrethroids: cis- and trans-3-(2,2-dichlorovinyl)-2,2-dimethylcyclopropane-1-carboxylic acids (cis-
Xiaojuan Qi et al.
International journal of hygiene and environmental health, 215(5), 487-495 (2012-01-06)
Pyrethroid pesticides are widely used throughout the world in agriculture to protect crops and in public health to control diseases. Of particular concern is exposure of pregnant women and their fetuses because little is known about the potential developmental hazards

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service