Skip to Content
Merck
All Photos(1)

Key Documents

31229

Supelco

Terbuthylazin-desethyl

PESTANAL®, analytical standard

Synonym(s):

2-Amino-4-tert-butylamino-6-chloro-1,3,5-triazine, Desethylterbutylazine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H12ClN5
CAS Number:
Molecular Weight:
201.66
Beilstein:
514392
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
clinical
environmental

format

neat

SMILES string

CC(C)(C)Nc1nc(N)nc(Cl)n1

InChI

1S/C7H12ClN5/c1-7(2,3)13-6-11-4(8)10-5(9)12-6/h1-3H3,(H3,9,10,11,12,13)

InChI key

LMKQNTMFZLAJDV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Terbuthylazin-desethyl is a triazine herbicide used for the control and elimination of weeds.[1]

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Terbuthylazin-desethyl may be used as a reference standard for the determination of terbuthylazin-desethyl in water samples using dispersed single-walled carbon nanohorns as sorbent and gas chromatography-mass spectrometry as instrumental technique.[1]

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dispersive micro solid-phase extraction of triazines from waters using oxidized single-walled carbon nanohorns as sorbent.
Jimenez-Soto JM, et al.
Journal of Chromatography A, 1245, 17-23 (2012)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service