Skip to Content
Merck
All Photos(1)

Key Documents

RIE156317687

Rieke Metals, Inc.

cis-2-(2-thiomethylbenzoyl)cyclopentane-1-carboxylic acid

All chiral compounds contain a mixture of enantiomers i.e. not chirally pure.

Sign Into View Organizational & Contract Pricing


About This Item

Molecular Weight:
264.35
MDL number:

Iupac Name

(1R,2S)-2-[2-(methylsulfanyl)benzoyl]cyclopentanecarboxylic acid

Inchi Code

1S/C14H16O3S/c1-18-12-8-3-2-5-11(12)13(15)9-6-4-7-10(9)14(16)17/h2-3,5,8-10H,4,6-7H2,1H3,(H,16,17)/t9-,10+/m0/s1

InChI key

ZUWIBRLZGCPRKH-VHSXEESVSA-N

Purity

95%

Country of Origin

US

Physical Form

White to Off-white solid

Product Link

https://riekemetals.com/products-services/catnum/18009-14

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service