Skip to Content
Merck
All Photos(2)

Key Documents

SYMBDSR02

Millipore

Steritest® Symbio Drain Tray Support O-rings

Accessory to streamline the workflow and increase safety during sterility testing

Synonym(s):

O-rings for drain tray support

Sign Into View Organizational & Contract Pricing

Select a Size

5 PKG
356,00 kr.

356,00 kr.


Please contact Customer Service for Availability

Request more information

Select a Size

Change View
5 PKG
356,00 kr.

About This Item

UNSPSC Code:
23151818
NACRES:
NB.79

356,00 kr.


Please contact Customer Service for Availability

Request more information

material

EPDM rubber O-ring

Agency

complies with EP (2.6.1)
complies with JP (4.06)
complies with USP (71)

packaging

pkg of 1 unit

manufacturer/tradename

Steritest®

application(s)

pharmaceutical
sterility testing

compatibility

for use with Steritest® Symbio Pump

shipped in

ambient

General description

The drain tray support O-rings is an accessory for the Steritest® Symbio pumps. Steritest® Symbio Pumps allow reliable, faster, and easy sterility testing in laminar flow hoods, isolators, and cleanrooms. The O-rings assure safe and leak-free operation in the drain tray as well as safe and easy handling of the canisters

Application

The drain tray support O-rings is an accessory for the Steritest® Symbio pumps, that is used in streamlining the workflow and increasing safety during sterility testing.

Legal Information

STERITEST is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service