Skip to Content
Merck
All Photos(1)

Key Documents

MABN359

Sigma-Aldrich

Anti-SLC22A3 Antibody, clone 4H6.1

clone 4H6.1, from mouse

Synonym(s):

Solute carrier family 22 member 3, Extraneuronal monoamine transporter, EMT, Organic cation transporter 3, SLC22A3

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
2.360,00 kr.

2.360,00 kr.


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 μG
2.360,00 kr.

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.43

2.360,00 kr.


Please contact Customer Service for Availability

Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

4H6.1, monoclonal

species reactivity

human

technique(s)

western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

target post-translational modification

unmodified

Gene Information

human ... SLC22A3(6581)

General description

Solute carrier family 22 member 3 (UniProt O75751; also known as EMT, Extraneuronal monoamine transporterorganic cation transporter 3, Organic cation transporter 3) is encoded by the SLC22A3 (also known as EMTH, OCT3) gene (Gene ID 6581) in human. Solute carrier family 22 member 3 is a multi-transmembrane protein that mediates the outward transport and clearance of endogenous small organic cations as well as a wide array of drugs and environmental toxins. Known substrates for this transporter include histamine, serotonin, norepinephrine, dopamine and MPP+.

Immunogen

Epitope: C-terminal
GST-tagged recombinant protein corresponding to the C-terminal of human SLC22A3.

Application

Anti-SLC22A3 Antibody, clone 4H6.1, Cat. No. MABN359, is a highly specific mouse monoclonal antibody, that targets SLC22A3 and has been tested in Western Blotting.
Research Category
Neuroscience
Research Sub Category
Developmental Signaling

Quality

Evaluated by Western Blotting in human brain tissue lysate.

Western Blotting Analysis: 2.0 µg/mL of this antibody detected SLC22A3 in 10 µg of human brain tissue lysate.

Target description

~70 kDa observed. Due to glycosylation, the target band appears larger than the calculated molecular weight of 61.3 kDa

Linkage

Replaces: MABN1039

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service