Skip to Content
Merck
All Photos(1)

Key Documents

MAB88079

Sigma-Aldrich

Anti-Heat Shock Factor II Antibody, clone 3E2

clone 3E2, Chemicon®, from rat

Synonym(s):

HSFII

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
2.910,00 kr.

2.910,00 kr.


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
50 μG
2.910,00 kr.

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.43

2.910,00 kr.


Please contact Customer Service for Availability

Request a Bulk Order

biological source

rat

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

3E2, monoclonal

species reactivity

human, monkey, mouse

manufacturer/tradename

Chemicon®

technique(s)

ELISA: suitable
immunofluorescence: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG2a

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... HSF2(3298)

Specificity

MAB88079 recognizes a protein of 70 kDa, identified as Heat Shock Factor 2 (HSF2). The synthesis of heat shock proteins is dependent on transcriptional and post-transcriptional control in eukaryotic cells. The regulation of heat shock gene transcription is mediated by the transcriptional activator, Heat Shock Factor (HSF), which binds to heat shock response elements (HSEs). At least three HSFs have been isolated from the human, mouse, chicken, and tomato genomes and an additional factor, HSF4 has been identified in humans. HSFs purified from yeast, Drosphila, and human have different molecular weights and do not cross-react immunologically. These factors are activated by distinct stimuli. HSF1 responds to cellular stress signals such as heat, heavy metals, and oxidative reagents whereas, the HSF2 is activated during differentiation. some laboratories have reported that the constitutive, non-DNA binding, monomeric form of inactive HSF1 is located in the cytoplasm. Upon activation, HSF1 is trimerized and is localized in nucleus where it binds DNA. the phosphorylation of HSF1 may be necessary for maximal transcription of heat shock genes.

Immunogen

Full length recombinant mouse HSF2 proteins (aa 1-517)

Application

Anti-Heat Shock Factor II Antibody, clone 3E2 is an antibody against Heat Shock Factor II for use in ELISA, IF, IP & WB.
Immunofluorescence Immunohistology (predominantly cytoplasmic with some nuclear). Immunoprecipitation: 2 μg/mg protein lysate. Western blot. 1:200-1:400 ELISA

Optimal working dilutions must be determined by end user.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Transcription Factors

Physical form

Format: Purified
Purified immunoglobulin. Liquid containing 10 mM PBS, pH 7.4, with 0.2% BSA and 15 mM sodium azide.

Storage and Stability

Maintain refrigerated at 2-8°C in undiluted aliquots for up to 12 months.

Analysis Note

Control
POSITIVE CONTROL:

NIH-3T3 cells

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nitzan Levy et al.
Endocrinology, 148(7), 3449-3458 (2007-03-31)
Estrogen receptors (ERs) regulate transcription by interacting with regulatory elements in target genes. However, known ER regulatory elements cannot explain the expression profiles of genes activated by estradiol (E2) and selective estrogen receptor modulators (SERMs). We previously showed that the
Linda Pesonen et al.
Cell stress & chaperones, 26(5), 819-833 (2021-08-01)
Cancer cells rely on heat shock proteins (HSPs) for growth and survival. Especially HSP90 has multiple client proteins and plays a critical role in malignant transformation, and therefore different types of HSP90 inhibitors are being developed. The bioactive natural compound
Marek A Budzyński et al.
Molecular and cellular biology, 35(14), 2530-2540 (2015-05-13)
In mammals the stress-inducible expression of genes encoding heat shock proteins is under the control of the heat shock transcription factor 1 (HSF1). Activation of HSF1 is a multistep process, involving trimerization, acquisition of DNA-binding and transcriptional activities, which coincide

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service