Skip to Content
Merck
All Photos(1)

Documents

8.41493

Sigma-Aldrich

2-Hydroxy-5-methoxybenzaldehyde

for synthesis

Synonym(s):

2-Hydroxy-5-methoxybenzaldehyde, 5-Methoxysalicylaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H8O3
CAS Number:
Molecular Weight:
152.15
MDL number:
UNSPSC Code:
12352114
EC Index Number:
211-589-0

Assay

≥98.0% (GC)

form

liquid

bp

100-102 °C/2.67 hPa

mp

4 °C

transition temp

flash point >110 °C

density

1.22 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C8H8O3/c1-11-7-2-3-8(10)6(4-7)5-9/h2-5,10H,1H3

InChI key

FZHSPPYCNDYIKD-UHFFFAOYSA-N

Application

2-Hydroxy-5-methoxybenzaldehyde for synthesis. CAS 672-13-9, molar mass 152.15 g/mol.

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.220 - 1.224
Identity (IR): passes test

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(C)

113 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service