Skip to Content
Merck
All Photos(1)

Key Documents

8.21017

Sigma-Aldrich

Diphenyl sulfide

for synthesis

Synonym(s):

Diphenyl sulfide, Phenyl sulfide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5)2S
CAS Number:
Molecular Weight:
186.27
MDL number:
UNSPSC Code:
12352113
EC Index Number:
205-371-4

vapor pressure

0.01 hPa ( 25 °C)

Assay

≥98.0% (GC)

form

liquid

potency

544 mg/kg LD50, oral (Rat)
12543 mg/kg LD50, skin (Rabbit)

bp

296 °C/1013 hPa

mp

-40 °C

transition temp

flash point >110 °C

density

1.11 g/cm3 at 20 °C

storage temp.

2-30°C

InChI

1S/C12H10S/c1-3-7-11(8-4-1)13-12-9-5-2-6-10-12/h1-10H

InChI key

LTYMSROWYAPPGB-UHFFFAOYSA-N

Analysis Note

Assay (GC, area%): ≥ 98.0 % (a/a)
Density (d 20 °C/ 4 °C): 1.112 - 1.114
Identity (IR): passes test

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service