Skip to Content
Merck
All Photos(1)

Key Documents

6.18418

Supelco

Silica gel

self-indicating coarse

Synonym(s):

Silica gel

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352309
Pricing and availability is not currently available.

bp

2230 °C

mp

>1600 °C

SMILES string

O=[Si]=O

InChI

1S/O2Si/c1-3-2

InChI key

VYPSYNLAJGMNEJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

  • Chemical Investigation on the Volatile Part of the CO(2) Supercritical Fluid Extract of Infected Aquilaria sinensis (Chinese Agarwood).: This article presents the use of silica gel in the purification process during the chemical analysis of volatile compounds from the CO2 supercritical fluid extract of infected Chinese agarwood. The study showcases the pivotal role of silica gel in isolating and analyzing fragrant compounds which are valuable in perfumery and traditional medicine (Mladenovic et al., 2024).
  • One-Pot Preparation of Mixed-Mode Reversed-Phase Anion-Exchange Silica Sorbent and its Application in the Detection of Cyclopiazonic Acid in Feeds and Agricultural Products.: This research demonstrates the development of a mixed-mode reversed-phase anion-exchange silica sorbent. Utilizing silica gel, this study highlights its application in the detection of mycotoxins, providing a robust method for ensuring the safety and quality of agricultural products (Hu et al., 2024).
  • Site-specific immobilization of Cysteinyl leukotriene receptor 1 through enzymatic DNA-protein conjugation strategy for lead screening.: This paper explores a novel approach to immobilize proteins on silica surfaces, enhancing the efficiency of biochemical assays. The application of silica gel in this context supports advanced biochemical research and the development of therapeutic agents, showcasing its critical role in medical research and diagnostics (Wen et al., 2024).
  • The Molecular Mechanism of FABP4 Inhibition Effects of GAS and 4-HBA in Gastrodia elata Blume Was Discussed Based on NMR and Molecular Docking.: In this study, silica gel plays a crucial role in the purification steps required for the structural elucidation of bioactive compounds from Gastrodia elata Blume. This research contributes to understanding the inhibition mechanisms of specific biomolecules, with implications for therapeutic applications (Yang et al., 2024).

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Carc. 1B

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

CAS No.

EU REACH Annex XVII (Restriction List)

CAS No.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service