Skip to Content
Merck
All Photos(1)

Key Documents

372770

Sigma-Aldrich

HBDDE

A cell-permeable protein kinase C (PKC) inhibitor that selectively inhibits PKC&alpha and PKCγ over PKCδ, βI, and βII isozymes in in vitro assays.

Synonym(s):

HBDDE, 2,2ʹ,3,3ʹ,4,4ʹ-Hexahydroxy-1,1ʹ-biphenyl-6,6ʹ-dimethanol Dimethyl Ether

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
2.040,00 kr.

2.040,00 kr.


Please contact Customer Service for Availability


Select a Size

Change View
1 MG
2.040,00 kr.

About This Item

Empirical Formula (Hill Notation):
C16H18O8
CAS Number:
Molecular Weight:
338.31
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

2.040,00 kr.


Please contact Customer Service for Availability

Quality Level

Assay

≥95% (TLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

orange

solubility

ethanol: 1 mg/mL
warm water: 1 mg/mL
DMSO: 5 mg/mL

shipped in

ambient

storage temp.

−20°C

SMILES string

O(Cc1c(c(c(c(c1)O)O)O)c2c(c(c(cc2COC)O)O)O)C

InChI

1S/C16H18O8/c1-23-5-7-3-9(17)13(19)15(21)11(7)12-8(6-24-2)4-10(18)14(20)16(12)22/h3-4,17-22H,5-6H2,1-2H3

InChI key

NEBCAMAQXZIVRE-UHFFFAOYSA-N

General description

A cell-permeable protein kinase C (PKC) inhibitor that selectively inhibits PKCα (IC50 = 43 µM) and PKCγ (IC50 = 50 µM) over PKCδ, βI, and βII isozymes in in vitro assays. Reported to induce apoptosis in rodent cerebellar granule neurons.
Protein kinase C (PKC) inhibitor. Selectively inhibits PKCα (IC50 = 43 µM) and PKCγ (IC50 = 50 µM) over PKCδ, βI and βII isozymes.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
PKCα
Product does not compete with ATP.
Reversible: no
Target IC50: 43 µM against PKCα

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Mathur, A., and Vallano, M.L. 2000. Biochem. Pharmacol.60, 809.
Kashiwada, Y., et al. 1994. J. Med. Chem. 37, 195.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Curtis B Read et al.
mBio, 15(4), e0029924-e0029924 (2024-02-28)
Many intracellular pathogens structurally disrupt the Golgi apparatus as an evolutionarily conserved promicrobial strategy. Yet, the host factors and signaling processes involved are often poorly understood, particularly for Anaplasma phagocytophilum, the agent of human granulocytic anaplasmosis. We found that A.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service