Skip to Content
Merck
All Photos(1)

Key Documents

14-700

Sigma-Aldrich

NEK11 Protein, active, 10 µg

Sign Into View Organizational & Contract Pricing

Select a Size

14-700
3.080,00 kr.

3.080,00 kr.


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
14-700
3.080,00 kr.

About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405

3.080,00 kr.


Please contact Customer Service for Availability

Request a Bulk Order

recombinant

expressed in baculovirus infected Sf21 cells

Quality Level

description

protein target NEK11

purified by

(Ni2+/NTA-agarose)

technique(s)

activity assay: suitable

UniProt accession no.

Gene Information

human ... NEK11(79858)

General description

FUNCTION: SwissProt: Q8NG66 # Possible role in S-phase checkpoint mechanism.
COFACTOR: Manganese or magnesium.
SIZE: 645 amino acids; 74162 Da
SUBCELLULAR LOCATION: Nucleus. Nucleus, nucleolus. Note=Nuclear during interphase but moves to the polar microtubules during prometaphase and metaphase. Accumulates in the nucleolus in G1/S- arrested cells.
TISSUE SPECIFICITY: Poorly expressed in cerebellum, trachea, lung, appendix, and uterus.
DEVELOPMENTAL STAGE: Predominantly expressed at S/G2/M phase.
PTM: Phosphorylated.
SIMILARITY: SwissProt: Q8NG66 ## Belongs to the protein kinase superfamily. Ser/Thr protein kinase family. NIMA subfamily. & Contains 1 protein kinase domain.

Quality

routinely evaluated by phosphorylation of MBP (Catalog # 13-104)

Physical properties

59.3kDa

Storage and Stability

6 months at -70°C from date of shipment

Analysis Note

For Specific Activity data, refer to the Certificate of Analysis for individual lots of this enzyme.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service