ENAH9453D7F4
Enamine
tert-butyl 4-[4-methyl-6-(trifluoromethanesulfonyloxy)pyrimidin-2-yl]piperidine-1-carboxylate
About This Item
Iupac Name
tert-butyl 4-(4-methyl-6-(((trifluoromethyl)sulfonyl)oxy)pyrimidin-2-yl)piperidine-1-carboxylate
Inchi Code
1S/C16H22F3N3O5S/c1-10-9-12(27-28(24,25)16(17,18)19)21-13(20-10)11-5-7-22(8-6-11)14(23)26-15(2,3)4/h9,11H,5-8H2,1-4H3
InChI key
QUMWQSMSVXZKLH-UHFFFAOYSA-N
Purity
Country of Origin
Product Link
MSDS
https://www.enaminestore.com/catalog/EN300-26981324
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service