Skip to Content
Merck
All Photos(1)

Key Documents

ENAH93E744BF

Enamine

8,8-difluoro-2-(iodomethyl)-1-oxaspiro[4.5]decane

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
3.272,25 kr.
100 MG
4.705,50 kr.
250 MG
6.577,50 kr.
500 MG
10.165,50 kr.
1 G
12.934,50 kr.
2.5 G
25.034,25 kr.

Vendor SKU: EN300-378259-50MG

3.272,25 kr.


Ships in 14 days - from Aldrich Partner.


Select a Size

Change View
50 MG
3.272,25 kr.
100 MG
4.705,50 kr.
250 MG
6.577,50 kr.
500 MG
10.165,50 kr.
1 G
12.934,50 kr.
2.5 G
25.034,25 kr.

About This Item

CAS Number:
Molecular Weight:
316.13
MDL number:

Vendor SKU: EN300-378259-50MG

3.272,25 kr.


Ships in 14 days - from Aldrich Partner.

Iupac Name

8,8-difluoro-2-(iodomethyl)-1-oxaspiro[4.5]decane

Inchi Code

1S/C10H15F2IO/c11-10(12)5-3-9(4-6-10)2-1-8(7-13)14-9/h8H,1-7H2

InChI key

PYMNQGDQCXEDIG-UHFFFAOYSA-N

Purity

95

Country of Origin

UA

Product Link

https://www.enaminestore.com/catalog/EN300-378259

MSDS

https://www.enaminestore.com/catalog/EN300-378259


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service