ENAH3045398D
Enamine
2-chloro-1-(2,5-dichlorophenyl)ethan-1-one
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Iupac Name
2-chloro-1-(2,5-dichlorophenyl)ethanone
Inchi Code
1S/C8H5Cl3O/c9-4-8(12)6-3-5(10)1-2-7(6)11/h1-3H,4H2
InChI key
CXEVBWANBXAQSY-UHFFFAOYSA-N
Country of Origin
UA
Product Link
https://www.enaminestore.com/catalog/EN300-08649
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service