Skip to Content
Merck
All Photos(1)

Key Documents

ENAH11E4156D

Enamine

3-ethynyl-1,1-dimethoxycyclobutane

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
140.18
MDL number:
Pricing and availability is not currently available. Contact Technical Service

Iupac Name

3-ethynyl-1,1-dimethoxycyclobutane

Inchi Code

1S/C8H12O2/c1-4-7-5-8(6-7,9-2)10-3/h1,7H,5-6H2,2-3H3

InChI key

NQRJMOLVGHCUOP-UHFFFAOYSA-N

storage temp.

4

Purity

95

Country of Origin

UA

Shipping Temperature

Normal

Physical Form

liquid

Product Link

https://www.enaminestore.com/catalog/EN300-2008054

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service