ENA388699877
Enamine
6-(2-methoxyethoxy)pyridine-3-carbonitrile
Select a Size
Vendor SKU: EN300-123159-100MG
591,00 kr.
Select a Size
About This Item
Vendor SKU: EN300-123159-100MG
591,00 kr.
Iupac Name
6-(2-methoxyethoxy)nicotinonitrile
Inchi Code
1S/C9H10N2O2/c1-12-4-5-13-9-3-2-8(6-10)7-11-9/h2-3,7H,4-5H2,1H3
InChI key
JNFWYSQNFRCPTR-UHFFFAOYSA-N
storage temp.
Purity
Country of Origin
Shipping Temperature
Physical Form
Product Link
Signal Word
Danger
Hazard Statements
MSDS
https://www.enaminestore.com/catalog/EN300-123159
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service