Skip to Content
Merck
All Photos(1)

Key Documents

CH4102153739

ChemBridge Corporation

3-chloro-N-(2-isopropylphenyl)propanamide

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
225.72
MDL number:

Pricing and availability is not currently available.

Iupac Name

3-chloro-N-(2-isopropylphenyl)propanamide

Inchi Code

1S/C12H16ClNO/c1-9(2)10-5-3-4-6-11(10)14-12(15)7-8-13/h3-6,9H,7-8H2,1-2H3,(H,14,15)

InChI key

DDVNYPWZRZDETF-UHFFFAOYSA-N

storage temp.

RT

Purity

95%

Country of Origin

KZ

Shipping Temperature

RT

Physical Form

solid

Product Link

https://www.hit2lead.com/building-blocks/7285066

MSDS

https://www.hit2lead.com/sds/7285066_sds.pdf


  • Choose from one of the most recent versions:

    Certificates of Analysis (COA)

    Lot/Batch Number

    Sorry, we don't have COAs for this product available online at this time.

    If you need assistance, please contact Customer Support.

    Already Own This Product?

    Find documentation for the products that you have recently purchased in the Document Library.

    Visit the Document Library

    Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

    Contact Technical Service