Skip to Content
Merck
All Photos(1)

Key Documents

ATC359510157

Atlantic Research Chemicals Ltd

Methyl 3-[(4-Formylphenoxy)methyl]benzoate

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
652,80 kr.
100 MG
804,00 kr.
250 MG
1.056,00 kr.
500 MG
1.308,00 kr.
1 G
1.812,00 kr.
2.5 G
3.273,60 kr.

Vendor SKU: ES001160-50MG

652,80 kr.


Ships in 14 days - from Aldrich Partner.


Select a Size

Change View
50 MG
652,80 kr.
100 MG
804,00 kr.
250 MG
1.056,00 kr.
500 MG
1.308,00 kr.
1 G
1.812,00 kr.
2.5 G
3.273,60 kr.

About This Item

CAS Number:
Molecular Weight:
270.28
MDL number:

Vendor SKU: ES001160-50MG

652,80 kr.


Ships in 14 days - from Aldrich Partner.

Iupac Name

methyl 3-[(4-formylphenoxy)methyl]benzoate

Inchi Code

1S/C16H14O4/c1-19-16(18)14-4-2-3-13(9-14)11-20-15-7-5-12(10-17)6-8-15/h2-10H,11H2,1H3

InChI key

TUVRKWCXTIAMID-UHFFFAOYSA-N

Purity

95

Country of Origin

GB

Product Link

https://atlantic-chemicals.com/product/?code=ES001160

MSDS

https://atlantic-chemicals.com/product/?code=ES001160


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service