Skip to Content
Merck
All Photos(1)

Key Documents

ATE398813571

AstaTech, Inc.

5-AMINO-2-(PHENYLTHIO)BENZOIC ACID

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
245.3
MDL number:
Pricing and availability is not currently available. Contact Technical Service

Iupac Name

5-amino-2-(phenylsulfanyl)benzoic acid

Inchi Code

1S/C13H11NO2S/c14-9-6-7-12(11(8-9)13(15)16)17-10-4-2-1-3-5-10/h1-8H,14H2,(H,15,16)

InChI key

MKSHKRLNPIFOSW-UHFFFAOYSA-N

storage temp.

Room temperature

Purity

95%

Country of Origin

CN

Shipping Temperature

Normal

Physical Form

White to Yellow Solid

Product Link

http://astatechinc.com/CPSResult.php?CRNO=19982

MSDS

http://astatechinc.com/Asta_MSDS.php?Catalog=AB1111


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service