Skip to Content
Merck
All Photos(1)

Key Documents

APO455832648

Apollo Scientific Ltd

4-Methyl-1,3-oxazole-5-carbonitrile

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
108.1
MDL number:

mp

60

density

1.12

Iupac Name

4-methyl-1,3-oxazole-5-carbonitrile

Inchi Code

1S/C5H4N2O/c1-4-5(2-6)8-3-7-4/h3H,1H3

InChI key

JZSLPQXOLZCAME-UHFFFAOYSA-N

storage temp.

Ambient Temperature

Country of Origin

GB

Shipping Temperature

Ambient Temperature

Physical Form

Liquid

Product Link

https://store.apolloscientific.co.uk/product/4-methyl-13-oxazole-5-carbonitrile

Flash Point

162/763mm

Melting Point

60

density

1.12

Pictograms

Signal Word

Warning

Hazard Statements

Precautionary statements

MSDS

https://store.apolloscientific.co.uk/product/4-methyl-13-oxazole-5-carbonitrile


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service