Skip to Content
Merck
All Photos(3)

Key Documents

T82007

Sigma-Aldrich

1,3,5-Triphenylbenzene

97%

Synonym(s):

1,3,5-Triphenylbenzene, 5′-Phenyl-1,1′:3′,1′′-terphenyl, 5′-Phenyl-m-terphenyl

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5)3C6H3
CAS Number:
Molecular Weight:
306.40
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

bp

460 °C (lit.)

mp

172-174 °C (lit.)

SMILES string

c1ccc(cc1)-c2cc(cc(c2)-c3ccccc3)-c4ccccc4

InChI

1S/C24H18/c1-4-10-19(11-5-1)22-16-23(20-12-6-2-7-13-20)18-24(17-22)21-14-8-3-9-15-21/h1-18H

InChI key

SXWIAEOZZQADEY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Daria M Arkhipova et al.
Nanomaterials (Basel, Switzerland), 10(12) (2020-12-16)
A new family of sterically hindered alkyl(tri-tert-butyl) phosphonium salts (n-CnH2n+1 with n = 2, 4, 6, 8, 10, 12, 14, 16, 18, 20) was synthesized and evaluated as stabilizers for the formation of palladium nanoparticles (PdNPs), and the prepared PdNPs
Biljana D Škrbić et al.
Environmental science and pollution research international, 24(19), 16148-16159 (2017-05-26)
Contents of 16 polycyclic aromatic hydrocarbons were analyzed in 30 soil samples from 15 locations in Novi Sad, Serbia, assessing for the first time the corresponding health risks in the Serbian urban zone. Total concentrations were in the range of
Perman Jorayev et al.
ChemSusChem, 13(23), 6433-6441 (2020-10-16)
Effective removal of excess greenhouse gas CO2 necessitates new adsorbents that can overcome the shortcomings of the current capture methods. To achieve that, porous materials are often modified post-synthetically with reactive amine functionalities but suffer from significant surface area losses.
Jian Chen et al.
Chemistry, an Asian journal, 12(17), 2291-2298 (2017-06-21)
CO
Vojtěch Štejfa et al.
Chemphyschem : a European journal of chemical physics and physical chemistry, 21(9), 938-951 (2020-04-02)
This work presents a new Knudsen effusion apparatus employing continuous monitoring of sample deposition using a quartz-crystal microbalance sensor with internal calibration by gravimetric determination of the sample mass loss. The apparatus was tested with anthracene and 1,3,5-triphenylbenzene and subsequently

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service