CLS3953312EC
Pyrex® gas-dispersion tube
pore size 70-220 μm
Synonym(s):
Corning® 39533
All Photos(1)
About This Item
Recommended Products
manufacturer/tradename
Corning 39533-12EC
pore size
70-220 μm
General description
With 12 mm O.D. fritted cylinder, 250 mm L × 8 mm diam. stem. Used to disperse gas in liquids. Size of bubbles depends on porosity. Size permits insertion through small openings.
Legal Information
Corning is a registered trademark of Corning, Inc.
PYREX is a registered trademark of Corning, Inc.