Skip to Content
Merck
All Photos(1)

Documents

771554

Sigma-Aldrich

9,9′-(9,9-Dihexyl-9H-fluorene-2,7-diyl)bis(3,6-di-tert-butylcarbazole)

Synonym(s):

9,9′-(9,9-Dihexyl-9H-fluorene-2,7-diyl)bis(3,6-di-tert-butyl-9H-carbazole), TBCF

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C65H80N2
Molecular Weight:
889.34
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

form

powder

mp

201-207 °C

λmax

350 nm in dichloromethane

fluorescence

λem 375 nm in chloroform

SMILES string

CCCCCCC1(CCCCCC)c2cc(ccc2-c3ccc(cc13)-n4c5ccc(cc5c6cc(ccc46)C(C)(C)C)C(C)(C)C)-n7c8ccc(cc8c9cc(ccc79)C(C)(C)C)C(C)(C)C

InChI

1S/C65H80N2/c1-15-17-19-21-35-65(36-22-20-18-16-2)55-41-47(66-57-31-23-43(61(3,4)5)37-51(57)52-38-44(62(6,7)8)24-32-58(52)66)27-29-49(55)50-30-28-48(42-56(50)65)67-59-33-25-45(63(9,10)11)39-53(59)54-40-46(64(12,13)14)26-34-60(54)67/h23-34,37-42H,15-22,35-36H2,1-14H3

InChI key

PRCXMWAOIXHYJA-UHFFFAOYSA-N

Application

Host materials for efficient solution-processed red and green phosphorescent devices

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Organic Light-emitting Diodes (OLEDs) are solid-state devices that transform electrical energy into light. OLEDs are considered the next generation technology for high-resolution flexible displays and solid state lighting, attracting intense scientific and industrial interest.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service