Skip to Content
Merck
All Photos(1)

Key Documents

617059

Sigma-Aldrich

Phenacetin-ethoxy-d5

98 atom % D

Synonym(s):

N-(4-Ethoxy-d5-phenyl)acetamide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CONHC6H4OCD2CD3
CAS Number:
Molecular Weight:
184.25
MDL number:
UNSPSC Code:
12352112
PubChem Substance ID:

Note: This product can be packaged on demand. For information on pricing, availability and packaging of custom sizes, please contact Stable Isotopes Customer Service

isotopic purity

98 atom % D

mp

134-136 °C (lit.)

mass shift

M+5

SMILES string

[2H]C([2H])([2H])C([2H])([2H])Oc1ccc(NC(C)=O)cc1

InChI

1S/C10H13NO2/c1-3-13-10-6-4-9(5-7-10)11-8(2)12/h4-7H,3H2,1-2H3,(H,11,12)/i1D3,3D2

InChI key

CPJSUEIXXCENMM-WNWXXORZSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Carc. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jerrie S Refuerzo et al.
Scientific reports, 6, 34710-34710 (2016-10-12)
Preterm labor caused by uterine contractions is a major contributor to neonatal morbidity and mortality. Treatment intended to reduce uterine contractions include tocolytic agents, such as indomethacin. Unfortunately, clinically used tocolytics are frequently inefficient and cross the placenta causing fetal

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service