Skip to Content
Merck
All Photos(1)

Key Documents

478997

Sigma-Aldrich

Methyl 3-isocyanatobenzoate

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
OCNC6H4CO2CH3
CAS Number:
Molecular Weight:
177.16
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

mp

35-38 °C (lit.)

storage temp.

2-8°C

SMILES string

COC(=O)c1cccc(c1)N=C=O

InChI

1S/C9H7NO3/c1-13-9(12)7-3-2-4-8(5-7)10-6-11/h2-5H,1H3

InChI key

WBGWGERFPSYHDT-UHFFFAOYSA-N

General description

Methyl 3-isocyanatobenzoate, also known as 3-(methoxycarbonyl)phenyl isocyanate, is an aryl isocyanate.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service