Skip to Content
Merck
All Photos(1)

Key Documents

229180

Sigma-Aldrich

(+)-Diisopropyl L-tartrate

98%, optical purity ee: 99% (GLC)

Synonym(s):

L-(+)-Tartaric acid diisopropyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[-CH(OH)CO2CH(CH3)2]2
CAS Number:
Molecular Weight:
234.25
Beilstein:
1727863
EC Number:
MDL number:
UNSPSC Code:
12352108
eCl@ss:
39021705
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

optical activity

[α]24/D +17°, neat

optical purity

ee: 99% (GLC)

refractive index

n20/D 1.439 (lit.)

bp

152 °C/12 mmHg (lit.)

density

1.114 g/mL at 25 °C (lit.)

SMILES string

CC(C)OC(=O)[C@H](O)[C@@H](O)C(=O)OC(C)C

InChI

1S/C10H18O6/c1-5(2)15-9(13)7(11)8(12)10(14)16-6(3)4/h5-8,11-12H,1-4H3/t7-,8-/m1/s1

InChI key

XEBCWEDRGPSHQH-HTQZYQBOSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Both D- and L-forms are reagents for kinetic resolution of racemic allylic alcohols and α-furfuryl amides by enantioselective epoxidation.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Journal of Organic Chemistry, 59, 1444-1444 (1994)
Aldrichimica Acta, 18, 53-53 (1985)
Tetrahedron, 49, 2641-2641 (1993)
Journal of the American Chemical Society, 115, 12208-12208 (1993)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service