Skip to Content
Merck
All Photos(1)

Key Documents

222267

Sigma-Aldrich

2-(tert-Butyl)anthracene

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H18
CAS Number:
Molecular Weight:
234.34
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

98%

mp

146-148 °C (lit.)

SMILES string

CC(C)(C)c1ccc2cc3ccccc3cc2c1

InChI

1S/C18H18/c1-18(2,3)17-9-8-15-10-13-6-4-5-7-14(13)11-16(15)12-17/h4-12H,1-3H3

InChI key

WBPXZSIKOVBSAS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2-(tert-Butyl)anthracene may be used in chemical synthesis studies.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service