Skip to Content
Merck
All Photos(1)

Key Documents

ADVH14DBE726

AChemBlock

N-(3-bromophenyl)cyclohexanecarboxamide

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
2.424,90 kr.
25 G
5.593,95 kr.

Vendor SKU: P51049-5G

2.424,90 kr.


Ships in 21 days - from Aldrich Partner.


Select a Size

Change View
5 G
2.424,90 kr.
25 G
5.593,95 kr.

About This Item

CAS Number:
Molecular Weight:
282.18
MDL number:

Vendor SKU: P51049-5G

2.424,90 kr.


Ships in 21 days - from Aldrich Partner.

Iupac Name

N-(3-bromophenyl)cyclohexanecarboxamide

Inchi Code

1S/C13H16BrNO/c14-11-7-4-8-12(9-11)15-13(16)10-5-2-1-3-6-10/h4,7-10H,1-3,5-6H2,(H,15,16)

InChI key

CRWRDUYMNRPRFM-UHFFFAOYSA-N

storage temp.

Store at Room Tem.

Purity

95%

Country of Origin

CN

Product Link

https://www.achemblock.com/p51049-n-3-bromophenyl-cyclohexanecarboxamide.html

Synonym

n-(3-bromophenyl)cyclohexanecarboxamide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary statements

MSDS

https://www.achemblock.com/p51049-n-3-bromophenyl-cyclohexanecarboxamide.html


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service