Merck
Interferon dimers: IFN-PEG-IFN.
Journal of drug targeting (2017-08-19)
Annabelle Herrington-Symes, Ji-Won Choi, Steve Brocchini
ABSTRACT
Increasingly complex proteins can be made by a recombinant chemical approach where proteins that can be made easily can be combined by site-specific chemical conjugation to form multifunctional or more active protein therapeutics. Protein dimers may display increased avidity for cell surface receptors. The increased size of protein dimers may also increase circulation times. Cytokines bind to cell surface receptors that dimerise, so much of the solvent accessible surface of a cytokine is involved in binding to its target. Interferon (IFN) homo-dimers (IFN-PEG-IFN) were prepared by two methods: site-specific bis-alkylation conjugation of PEG to the two thiols of a native disulphide or to two imidazoles on a histidine tag of two His
Sigma-Aldrich
Methyl violet 2B, certified by the BSC