Merck
  • Nongenomic actions of bile acids. Synthesis and preliminary characterization of 23- and 6,23-alkyl-substituted bile acid derivatives as selective modulators for the G-protein coupled receptor TGR5.
Nongenomic actions of bile acids. Synthesis and preliminary characterization of 23- and 6,23-alkyl-substituted bile acid derivatives as selective modulators for the G-protein coupled receptor TGR5.
Journal of medicinal chemistry (2007-08-10)
Roberto Pellicciari, Hiroyuki Sato, Antimo Gioiello, Gabriele Costantino, Antonio Macchiarulo, Bahman M Sadeghpour, Gianluca Giorgi, Kristina Schoonjans, Johan Auwerx
ABSTRACT
23-Alkyl-substituted and 6,23-alkyl-disubstituted derivatives of chenodeoxycholic acid are identified as potent and selective agonists of TGR5, a G-protein coupled receptor for bile acids (BAs). In particular, we show that methylation at the C-23(S) position of natural BAs confers a marked selectivity for TGR5 over FXR, while the 6alpha-alkyl substitution increases the potency at both receptors. The present results allow for the first time a pharmacological differentiation of genomic versus nongenomic effects mediated by BA derivatives.
Sigma-Aldrich
Chenodeoxycholic acid
Sigma-Aldrich
Cholic acid, from bovine and/or ovine, ≥98%