Merck
  • Highly sensitive detection of cancer cells with an electrochemical cytosensor based on boronic acid functional polythiophene.
Highly sensitive detection of cancer cells with an electrochemical cytosensor based on boronic acid functional polythiophene.
Biosensors & bioelectronics (2016-11-21)
Muamer Dervisevic, Mehmet Senel, Tugba Sagir, Sevim Isik
ABSTRACT
The detection of cancer cells through important molecular recognition target such as sialic acid is significant for the clinical diagnosis and treatment. There are many electrochemical cytosensors developed for cancer cells detection but most of them have complicated fabrication processes which results in poor reproducibility and reliability. In this study, a simple, low-cost, and highly sensitive electrochemical cytosensor was designed based on boronic acid-functionalized polythiophene. In cytosensors fabrication simple single-step procedure was used which includes coating pencil graphite electrode (PGE) by means of electro-polymerization of 3-Thienyl boronic acid and Thiophen. Electrochemical impedance spectroscopy and cyclic voltammetry were used as an analytical methods to optimize and measure analytical performances of PGE/P(TBA
Sigma-Aldrich
Thiophene, ≥99%
Sigma-Aldrich
3-Thienylboronic acid, ≥95.0%