Merck
  • Reversibly pH-responsive polyurethane membranes for on-demand intravaginal drug delivery.
Reversibly pH-responsive polyurethane membranes for on-demand intravaginal drug delivery.
Acta biomaterialia (2016-11-05)
Seungil Kim, Yufei Chen, Emmanuel A Ho, Song Liu
ABSTRACT
To provide better protection for women against sexually transmitted infections, on-demand intravaginal drug delivery was attempted by synthesizing reversibly pH-sensitive polyether-polyurethane copolymers using poly(ethylene glycol) (PEG) and 1,4-bis(2-hydroxyethyl)piperazine (HEP). Chemical structure and thermo-characteristics of the synthesized polyurethanes were confirmed by attenuated total reflectance-Fourier transform infrared spectroscopy (ATR-FTIR), A reversible and sharp switch between "off" and "on" drug release is achieved for the first time through new pH-sensitive polyurethane membranes, which can serve as window membranes in reservoir-type intravaginal rings for on-demand drug delivery to prevent sexually transmitted infections (STIs). Close to zero drug release occurs at the normal vaginal pH (4.5) for minimal side effects. Drug release is only triggered by elevation of pH to 7 during heterosexual intercourse. The reversibly sharp and fast "on-and-off" switch arises from the creative incorporation of a pH-sensitive monomer in the soft segment of polyurethane. This polyurethane biomaterial holds great potential to better protect women who are generally at higher risk and are more vulnerable to STIs.
Sigma-Aldrich
1,4-Bis(2-hydroxyethyl)piperazine, 99%
Sigma-Aldrich
1,2-Dichloroethane, anhydrous, 99.8%